4-Bromobenzaldehyde ≥97.0%
Supplier: TCI America
|
Warning
|
B0549-25G
B0549-250G
TCB0549-025GEA
132.5
CAD
TCB0549-025G
TCB0549-250G
4-Bromobenzaldehyde ≥97.0%
4-Bromobenzaldehyde
CAS Number: 1122-91-4
MDL Number: MFCD00003377
Molecular Formula: C7H5BrO
Molecular Weight: 185.02
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 57
Flash Point (°C): 108
Formula:
C₇H₅BrO Boiling Pt: 125 °C Melting Pt: 57 °C Density: 1.849 g/cm³ (20 °C) Flash Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003377 CAS Number: 1122-91-4 EINECS: 214-365-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...