3-Bromoanisole ≥98.0%

Supplier: TCI America

B0545-250G B0545-25G
TCB0545-250GEA 161.16 CAD
TCB0545-250G TCB0545-025G
3-Bromoanisole ≥98.0%
3-Bromoanisole

CAS Number: 2398-37-0
MDL Number: MFCD00000081
Molecular Formula: C7H7BrO
Molecular Weight: 187.04
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 211
Flash Point (°C): 98
Specific Gravity (20/20): 1.49

Formula: C₇H₇BrO
Boiling Pt: 211 °C
Melting Pt: 2 °C
Density: 1.49 (20 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00000081
CAS Number: 2398-37-0
EINECS: 219-264-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=951772af1827ce40be603e77eeb9c83cc45c0d5d&vid=48c19e36e38b634ed096f5c57640dde3a7e71200&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR