3-Bromoaniline ≥98.0%

Supplier: TCI America

B0542-25G B0542-100G
TCB0542-025GEA 75.52 CAD
TCB0542-025G TCB0542-100G
3-Bromoaniline ≥98.0%
3-Bromoaniline

CAS Number: 591-19-5
MDL Number: MFCD00007757
Molecular Formula: C6H6BrN
Molecular Weight: 172.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 251
Flash Point (°C): 110
Freezing point (°C): 17
Specific Gravity (20/20): 1.58

Formula: C₆H₆BrN
Boiling Pt: 251 °C
Melting Pt: 16 °C
Density: 1.58 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007757
CAS Number: 591-19-5
EINECS: 209-704-4
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad4bafd76eff30d86621241f0af3928acd41da79&vid=ad4bafd76eff30d86621241f0af3928acd41da79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR