4'-Bromoacetophenone ≥98.0%

Supplier: TCI America
Warning

Synonyms: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone

B0537-25G B0537-500G
TCB0537-025GEA 53.72 CAD
TCB0537-025G TCB0537-500G
4'-Bromoacetophenone ≥98.0%
4'-Bromoacetophenone

CAS Number: 99-90-1
MDL Number: MFCD00000105
Molecular Formula: C8H7BrO
Molecular Weight: 199.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 53
Flash Point (°C): 113

Formula: C₈H₇BrO
Boiling Pt: 256 °C
Melting Pt: 53 °C
Density: 1.736 g/cm³ (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000105
CAS Number: 99-90-1
EINECS: 202-799-3
UN: 2811
ADR: 6.1,II
Merck Index: 13,01387
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0762177456bb15031cf44947afe9798d7286cfe3&vid=0762177456bb15031cf44947afe9798d7286cfe3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR