3'-Bromoacetophenone ≥98.0%

Supplier: TCI America
Warning

Synonyms: m-Bromoacetophenone

B0536-5G B0536-25G
TCB0536-005GEA 26.86 CAD
TCB0536-005G TCB0536-025G
3'-Bromoacetophenone ≥98.0%
3'-Bromoacetophenone

CAS Number: 2142-63-4
MDL Number: MFCD00000083
Molecular Formula: C8H7BrO
Molecular Weight: 199.05
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 114
Freezing point (°C): 12
Specific Gravity (20/20): 1.50

Formula: C₈H₇BrO
MW: 199.05 g/mol
Boiling Pt: 136 °C
Density: 1.50 (20 °C)
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00000083
CAS Number: 2142-63-4
EINECS: 218-396-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b083d1724beff37f07ba2ec35ece52d1718d1ccb&vid=b083d1724beff37f07ba2ec35ece52d1718d1ccb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR