4'-Bromoacetanilide ≥98.0%
Supplier: TCI America
B0534-25G
B0534-100G
TCB0534-025GEA
110.6
CAD
TCB0534-025G
TCB0534-500G
TCB0534-100G
4'-Bromoacetanilide ≥98.0%
4'-Bromoacetanilide
CAS Number: 103-88-8
MDL Number: MFCD00000092
Molecular Formula: C8H8BrNO
Molecular Weight: 214.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 168
Formula:
H₃CC(O)NH(C₆H₄Br) Boiling Pt: 347 °C (1013 hPa) Melting Pt: 168 °C Density: 1.717 g/cm³ (20 °C) |
MDL Number:
MFCD00000092 CAS Number: 103-88-8 EINECS: 203-154-9 Merck Index: 13,01383 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...