1-Butylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

B0529-1G B0529-25G B0529-5G
TCB0529-001GEA 58.46 CAD
TCB0529-001G TCB0529-25G TCB0529-005G
1-Butylboronic acid (contains varying amounts of Anhydride)
1-Butylboronic acid

[for Esterification]
CAS Number: 4426-47-5
MDL Number: MFCD00002106
Molecular Formula: C4H11BO2
Molecular Weight: 101.94
Form: Crystal
Color: White
Melting point (°C): 90


Other Product Names / Synonyms: 1-Butaneboronic Acid

Formula: C₄H₁₁BO₂
Melting Pt: 90 °C
MDL Number: MFCD00002106
CAS Number: 4426-47-5
EINECS: 224-607-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=139092f7de31abdc80fb4a2a8864d060137d4108&vid=568a9d63fceaba0288f5e0b33e42d0f6c598bc27&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR