Tributyl borate ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B0518-500ML B0518-25ML B0518-100ML
TCB0518-500MLEA 181.7 CAD
TCB0518-500ML TCB0518-025ML TCB0518-100ML
Tributyl borate ≥99.0% (by titrimetric analysis)
Tributyl borate

CAS Number: 688-74-4
MDL Number: MFCD00009434
Molecular Formula: C12H27BO3
Molecular Weight: 230.16
Purity/Analysis Method: >99.0% (T)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 93
Specific Gravity (20/20): 0.86


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Boric Acid Tributyl Ester

Formula: C₁₂H₂₇BO₃
Boiling Pt: 230 °C
Melting Pt: –70 °C
Density: 0.86 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00009434
CAS Number: 688-74-4
EINECS: 211-706-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d29363d25e58460361a5e431f3ea03cc5c6fabc4&vid=d29363d25e58460361a5e431f3ea03cc5c6fabc4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR