Biuret ≥98.0% (by total nitrogen basis) (contains ca. 10% Triuret)
Supplier: TCI America
B0513-25G
B0513-500G
TCB0513-025GEA
143.78
CAD
TCB0513-025G
TCB0513-500G
Biuret ≥98.0% (by total nitrogen basis) (contains ca. 10% Triuret)
Biuret
CAS Number: 108-19-0
MDL Number: MFCD00007946
Molecular Formula: C2H5N3O2
Molecular Weight: 103.08
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 193
Other Product Names / Synonyms: Allophanamide
Formula:
C₂H₅N₃O₂ Melting Pt: 193 °C Density: 1.467 g/cm³ (10 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007946 CAS Number: 108-19-0 EINECS: 203-559-0 Merck Index: 13,01305 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...