1,4-Bis(5-phenyloxazol-2-yl)benzene ≥98.0%

Supplier: TCI America
Warning

Synonyms: POPOP, 1,4-Di(5-Phenyl-2-oxazolyl)benzene

B0509-25G B0509-5G B0509-1G
TCB0509-025GEA 376.04 CAD
TCB0509-025G TCB0509-5G TCB0509-001G
1,4-Bis(5-phenyloxazol-2-yl)benzene ≥98.0%
1,4-Bis(5-phenyloxazol-2-yl)benzene

[for scintillation spectrometry]
CAS Number: 1806-34-4
MDL Number: MFCD00005309
Molecular Formula: C24H16N2O2
Molecular Weight: 364.40
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 245


Product Categories: Analytical Science, Scintillators
Other Product Names / Synonyms: 1,4-Di(5-phenyl-2-oxazolyl)benzene, POPOP

Formula: C₂₄H₁₆N₂O₂
Melting Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00005309
CAS Number: 1806-34-4
EINECS: 217-304-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37e605fdd6d67fe77c8e3ad5ff0382f44fe2f4ce&vid=5036044557e5703aaf2be9b0d69169add4e0fa4c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR