Tetraethylene glycol dimethyl ether ≥98.0%
Supplier: TCI America
B0497-500ML
B0497-25ML
TCB0497-500MLEA
138.8
CAD
TCB0497-500ML
TCB0497-025ML
Tetraethylene glycol dimethyl ether ≥98.0%
Tetraethylene glycol dimethyl ether
CAS Number: 143-24-8
MDL Number: MFCD00008505
Molecular Formula: C10H22O5
Molecular Weight: 222.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 276
Flash Point (°C): 140
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: Bis[2-(2-methoxyethoxy)ethyl] Ether, Tetraglyme
Formula:
C₁₀H₂₂O₅ Boiling Pt: 276 °C Density: 1.01 (20 °C) Flash Pt: 140 °C |
MDL Number:
MFCD00008505 CAS Number: 143-24-8 EINECS: 205-594-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...