Triethylene glycol dimethyl ether ≥99.0% stabilized
Supplier: TCI America
Stabilization:
Stabilised with Stabilised
(stabilized with BHT)
CAS Number: 112-49-2
MDL Number: MFCD00008504
Molecular Formula: C8H18O4
Molecular Weight: 178.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 216
Melting point (°C): -45
Flash Point (°C): 108
Specific Gravity (20/20): 0.99
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: 1,2-Bis(2-methoxyethoxy)ethane, 2,5,8,11-Tetraoxadodecane, Triglyme
Formula:
C₈H₁₈O₄ Boiling Pt: 216 °C Melting Pt: –45 °C Density: 0.99 (20 °C) Flash Pt: 108 °C |
MDL Number:
MFCD00008504 CAS Number: 112-49-2 EINECS: 203-977-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...