Bis(2-ethoxyethyl)ether ≥98.0%

Supplier: TCI America

B0489-25ML B0489-500ML
TCB0489-025MLEA 31.6 CAD
TCB0489-025ML TCB0489-500ML
Bis(2-ethoxyethyl)ether ≥98.0%
Bis(2-ethoxyethyl) ether

CAS Number: 112-36-7
MDL Number: MFCD00009254
Molecular Formula: C8H18O3
Molecular Weight: 162.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 188
Melting point (°C): -44
Flash Point (°C): 71
Specific Gravity (20/20): 0.91


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Bis(2-ethoxyethyl) Ether, Diethyl Carbitol

Formula: C₈H₁₈O₃
Boiling Pt: 188 °C
Melting Pt: –44 °C
Density: 0.91 (20 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00009254
CAS Number: 112-36-7
EINECS: 203-963-7
Merck Index: 13,03145
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a134eeb8bc841df7ea77cdea019724a3d4cff84&vid=4a134eeb8bc841df7ea77cdea019724a3d4cff84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR