Bicine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine

B0484-500G B0484-25G B0484-100G
TCB0484-500GEA 417.12 CAD
TCB0484-500G TCB0484-025G TCB0484-100G
Bicine ≥99.0% (by titrimetric analysis)
Bicine

[Good's buffer component for biological research]
CAS Number: 150-25-4
MDL Number: MFCD00004295
Molecular Formula: C6H13NO4
Molecular Weight: 163.17
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 195


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: Bicine, N,N-Bis(2-hydroxyethyl)glycine, Diethanolglycine

Formula: C₆H₁₃NO₄
MW: 163.17 g/mol
Melting Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS Number: 150-25-4
EINECS: 205-755-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098679e466c6e52f6616ac73d11b8bc4b628af4c&vid=363c495e7ab227a637ab40ee554de2c546a352ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR