4,4'-Bis(dimethylamino)diphenylmethane ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Methylenebis(N,N-dimethylaniline), Bis[4-(dimethylamino)phenyl]methane
CAS Number: 101-61-1
MDL Number: MFCD00008317
Molecular Formula: C17H22N2
Molecular Weight: 254.38
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 390
Melting point (°C): 91
Flash Point (°C): 178
Product Categories: Materials Chemistry, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: Methane Base, 4,4'-Methylenebis(N,N-dimethylaniline), Tetra Base, 4,4'-Tetramethyldiaminodiphenylmethane
Formula:
C₁₇H₂₂N₂ MW: 254.38 g/mol Boiling Pt: 390 °C Melting Pt: 91 °C Flash Pt: 178 °C |
MDL Number:
MFCD00008317 CAS Number: 101-61-1 EINECS: 202-959-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...