4,4'-Bis(dimethylamino)benzophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

B0481-500G B0481-25G B0481-100G
TCB0481-500GEA 560.9 CAD
TCB0481-500G TCB0481-25G TCB0481-100G
4,4'-Bis(dimethylamino)benzophenone ≥98.0% (by GC, titration analysis)
4,4'-Bis(dimethylamino)benzophenone

CAS Number: 90-94-8
MDL Number: MFCD00008312
Molecular Formula: C17H20N2O
Molecular Weight: 268.36
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 360
Melting point (°C): 174
Flash Point (°C): 220


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: Michler's Ketone, N,N,N',N'-Tetramethyl-4,4'-diaminobenzophenone

Formula: C₁₇H₂₀N₂O
Boiling Pt: 360 °C
Melting Pt: 174 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00008312
CAS Number: 90-94-8
EINECS: 202-027-5
UN: 2811
ADR: 6.1,III
Merck Index: 12,06265
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2bd084b7efbb27eb705bad5e93639018bba03d4&vid=c2bd084b7efbb27eb705bad5e93639018bba03d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR