Methylthymol blue sodium salt ≥98.0%

Supplier: TCI America

Synonyms: Sodium 2,2',2'',2'''-((((1,1-dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetate, Methylthymol Blue tetrasodium salt

B0478-25G B0478-1G B0478-5G
TCB0478-025GEA 1472.62 CAD
TCB0478-025G TCB0478-001G TCB0478-5G
Methylthymol blue sodium salt ≥98.0%
Methylthymol Blue sodium salt

CAS Number: 1945-77-3
MDL Number: MFCD00148905
Molecular Formula: C37H44N2O13S
Molecular Weight: 844.75
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: 3,3'-Bis[N,N-bis(carboxymethyl)aminomethyl]thymolsulfonphthalein Sodium Salt

Formula: C₃₇H₄₀N₂Na₄O₁₃S
MW: 844.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00148905
CAS Number: 1945-77-3
EINECS: 217-743-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffabb83d04937d5072f18b99f65dde520f9e5c55&vid=ffabb83d04937d5072f18b99f65dde520f9e5c55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR