2-Phenylaniline ≥99.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B0467-25G
B0467-500G
B0467-100G
TCB0467-025GEA
189.76
CAD
TCB0467-025G
TCB0467-500G
TCB0467-100G
2-Phenylaniline ≥99.0% (by GC, titration analysis)
2-Phenylaniline
[for Sugar determination]
CAS Number: 90-41-5
MDL Number: MFCD00007701
Molecular Formula: C12H11N
Molecular Weight: 169.23
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 299
Melting point (°C): 51
Flash Point (°C): 153
Other Product Names / Synonyms: 2-Aminodiphenyl, 2-Biphenylamine
Formula:
C₁₂H₁₁N Boiling Pt: 299 °C Melting Pt: 51 °C Flash Pt: 153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007701 CAS Number: 90-41-5 EINECS: 201-990-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...