1,2-Diphenylethane ≥99.0%

Supplier: TCI America

Synonyms: Dibenzyl, Bibenzyl

B0459-25G B0459-500G B0459-100G
TCB0459-025GEA 76.33 CAD
TCB0459-025G TCB0459-500G TCB0459-100G
1,2-Diphenylethane ≥99.0%
1,2-Diphenylethane

CAS Number: 103-29-7
MDL Number: MFCD00004796
Molecular Formula: C14H14
Molecular Weight: 182.27
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 284
Melting point (°C): 52
Flash Point (°C): 129


Other Product Names / Synonyms: Bibenzyl, s-Diphenylethane

Formula: (C₆H₅)CH₂CH₂(C₆H₅)
MW: 182.27 g/mol
Boiling Pt: 284
Melting Pt: 52 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00004796
CAS Number: 103-29-7
EINECS: 203-096-4
Merck Index: 12,01245
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51cd6890890aff71d2194725abcb524caed2e393&vid=51cd6890890aff71d2194725abcb524caed2e393&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR