Betaine hydrochloride ≥98.0%

Supplier: TCI America

B0458-500G B0458-25G
TCB0458-500GEA 541.94 CAD
TCB0458-500G TCB0458-025G
Betaine hydrochloride ≥98.0%
Betaine hydrochloride

CAS Number: 590-46-5
MDL Number: MFCD00011903
Molecular Formula: C5H11NO2
Molecular Weight: 153.61
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₅H₁₂ClNO₂
MW: 153.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011903
CAS Number: 590-46-5
EINECS: 209-683-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69209c927e96e7c0550cdcfab3ec7da3bf5625b6&vid=69209c927e96e7c0550cdcfab3ec7da3bf5625b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR