Betaine, anhydrous ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(Trimethylammonio)acetateTrimethylglycineTMG

B0455-500G B0455-25G B0455-100G
TCB0455-025GEA 47.73 CAD
TCB0455-025G TCB0455-500G TCB0455-100G
Betaine, anhydrous ≥97.0% (by titrimetric analysis)
Betaine

CAS Number: 107-43-7
MDL Number: MFCD00012123
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 300

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00012123
CAS Number: 107-43-7
https://ca.vwr.com/stibo/web/std.lang.all/24/21/28932421.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR