Benzyltrimethylammonium hydroxide 40% in methanol
Supplier: TCI America
B0448-500ML
B0448-25ML
B0448-100ML
TCB0448-500MLEA
127.02
CAD
TCB0448-500ML
TCB0448-25ML
TCB0448-100ML
Benzyltrimethylammonium hydroxide 40% in methanol
Benzyltrimethylammonium hydroxide
CAS Number: 100-85-6
MDL Number: MFCD00008281
Molecular Formula: C10H17NO
Molecular Weight: 167.25
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 11
Specific Gravity (20/20): 0.92
Formula:
C₁₀H₁₇NO Density: 0.92 (20 °C) Flash Pt: 11 °C |
MDL Number:
MFCD00008281 CAS Number: 100-85-6 EINECS: 202-895-5 UN: 3286 ADR: 3,I |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...