Benzyltriethylammonium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B0444-25G B0444-500G
TCB0444-025GEA 38.33 CAD
TCB0444-025G TCB0444-500G
Benzyltriethylammonium chloride ≥98.0% (by titrimetric analysis)
Benzyltriethylammonium chloride

CAS Number: 56-37-1
MDL Number: MFCD00011824
Molecular Formula: C13H22ClN
Molecular Weight: 227.78
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 192
Flash Point (°C): 169


Other Product Names / Synonyms: BTEAC, TEBAC

Formula: C₁₃H₂₂ClN
Melting Pt: 192 °C
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00011824
CAS Number: 56-37-1
EINECS: 200-270-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b21b56b37fb0edc45c1c0ce83cb3080116f6938a&vid=b21b56b37fb0edc45c1c0ce83cb3080116f6938a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR