Deoxybenzoin ≥98.0%

Supplier: TCI America

B0435-250G B0435-25G
TCB0435-250GEA 222.78 CAD
TCB0435-250G TCB0435-025G
Deoxybenzoin ≥98.0%
Deoxybenzoin

CAS Number: 451-40-1
MDL Number: MFCD00003081
Molecular Formula: C14H12O
Molecular Weight: 196.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 322
Melting point (°C): 56
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Deoxybenzoin, 2-Phenylacetophenone

Formula: C₁₄H₁₂O
Boiling Pt: 322 °C
Melting Pt: 56 °C
Flash Pt: 110 °C
MDL Number: MFCD00003081
CAS Number: 451-40-1
EINECS: 207-193-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a28676540ccdf6a5178c45df97a77e486943d0aa&vid=a28676540ccdf6a5178c45df97a77e486943d0aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR