Diethyl benzylmalonate ≥98.0%

Supplier: TCI America

B0423-25ML B0423-500ML
TCB0423-025MLEA 142.2 CAD
TCB0423-025ML TCB0423-500ML
Diethyl benzylmalonate ≥98.0%
Diethyl benzylmalonate

CAS Number: 607-81-8
MDL Number: MFCD00009166
Molecular Formula: C14H18O4
Molecular Weight: 250.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 300
Flash Point (°C): 100
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: Benzylmalonic Acid Diethyl Ester

Formula: C₁₄H₁₈O₄
Boiling Pt: 300 °C
Density: 1.08 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00009166
CAS Number: 607-81-8
EINECS: 210-142-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df3d8a6fd62e7a02e02901018edb559d212ffa11&vid=df3d8a6fd62e7a02e02901018edb559d212ffa11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR