Benzylacetone ≥95.0%

Supplier: TCI America

Synonyms: 4-Phenyl-2-butanone, 4-Phenylbutan-2-one

B0405-500ML B0405-25ML
TCB0405-500MLEA 251.22 CAD
TCB0405-500ML TCB0405-025ML
Benzylacetone ≥95.0%
Benzylacetone

CAS Number: 2550-26-7
MDL Number: MFCD00008790
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 98
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Methyl Phenethyl Ketone, Methyl 2-Phenylethyl Ketone, Benzylacetone

Formula: (C₆H₅)CH₂CH₂C(O)CH₃
MW: 148.2 g/mol
Boiling Pt: 115 °C
Melting Pt: –13 °C
Density: 0.99 (20 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00008790
CAS Number: 2550-26-7
EINECS: 219-847-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1774cb7bf9a7e21599972fd2ac31503b676bc240&vid=6e863df7825f34ce4e8e5955930266d158c39424&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR