3,3'-Diamino diphenyl sulfone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B0394-25G B0394-500G
TCB0394-025GEA 44.37 CAD
TCB0394-025G TCB0394-500G
3,3'-Diamino diphenyl sulfone ≥98.0% (by titrimetric analysis)
3,3'-Diamino diphenyl sulphone

CAS Number: 599-61-1
MDL Number: MFCD00007792
Molecular Formula: C12H12N2O2S
Molecular Weight: 248.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 172


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Diphenyl Sulfones (for High-Performance Polymer Research)
Other Product Names / Synonyms: 3,3'-Diaminodiphenyl Sulfone, 3,3'-Sulfonyldianiline

Formula: C₁₂H₁₂N₂O₂S
Melting Pt: 172 °C
MDL Number: MFCD00007792
CAS Number: 599-61-1
EINECS: 209-967-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9069fcd7193128be84f213d4ea08894092d2df0&vid=e9069fcd7193128be84f213d4ea08894092d2df0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR