1-Bromoadamantane ≥99.0%
Supplier: TCI America
B0390-250G
B0390-25G
TCB0390-250GEA
194.97
CAD
TCB0390-250G
TCB0390-025G
1-Bromoadamantane ≥99.0%
1-Bromoadamantane
CAS Number: 768-90-1
MDL Number: MFCD00074721
Molecular Formula: C10H15Br
Molecular Weight: 215.13
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 119
Other Product Names / Synonyms: 1-Adamantyl Bromide
Formula:
C₁₀H₁₅Br Boiling Pt: 226 °C (1013 hPa) Melting Pt: 119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074721 CAS Number: 768-90-1 EINECS: 212-200-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...