Diethyl butylmalonate ≥99.0%

Supplier: TCI America

Synonyms: Butylmalonic acid diethyl ester, Butyl-1,3-propanedioic acid diethyl ester, Diethyl 2-butylmalonate

B0381-25ML B0381-500ML
TCB0381-025MLEA 34.76 CAD
TCB0381-025ML TCB0381-500ML
Diethyl butylmalonate ≥99.0%
Diethyl butylmalonate

CAS Number: 133-08-4
MDL Number: MFCD00009170
Molecular Formula: C11H20O4
Molecular Weight: 216.28
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 240
Flash Point (°C): 94
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Butylmalonic Acid Diethyl Ester

Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 240 °C
Density: 0.98 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00009170
CAS Number: 133-08-4
EINECS: 205-089-1
Merck Index: 13,01575
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96507d9d335426e75cda9d61841e94dbbb7c21de&vid=96507d9d335426e75cda9d61841e94dbbb7c21de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR