1-Butanesulfonic acid sodium salt ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B0379-25G
B0379-500G
TCB0379-025GEA
186.44
CAD
TCB0379-025G
TCB0379-500G
1-Butanesulfonic acid sodium salt ≥98.0% (by titrimetric analysis)
1-Butansulphonic acid sodium salt
CAS Number: 2386-54-1
MDL Number: MFCD00007540
Molecular Formula: C4H10O3S
Molecular Weight: 160.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 1-Butanesulfonic Acid Sodium Salt, Butylsulfonic Acid Sodium Salt
Formula:
C₄H₉NaO₃S Melting Pt: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007540 CAS Number: 2386-54-1 EINECS: 219-201-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...