3-Benzoylpyridine ≥99.0%

Supplier: TCI America

B0305-5G B0305-25G
TCB0305-5GEA 90.06 CAD
TCB0305-5G TCB0305-025G
3-Benzoylpyridine ≥99.0%
3-Benzoylpyridine

CAS Number: 5424-19-1
MDL Number: MFCD00006394
Molecular Formula: C12H9NO
Molecular Weight: 183.21
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 307
Melting point (°C): 39
Flash Point (°C): 150


Other Product Names / Synonyms: Phenyl 3-Pyridyl Ketone

Formula: C₁₂H₉NO
Boiling Pt: 307 °C
Melting Pt: 39 °C
Flash Pt: 150 °C
MDL Number: MFCD00006394
CAS Number: 5424-19-1
EINECS: 226-561-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58c2a1b1a2bba7b70c2b52a7c5bea443929d787a&vid=58c2a1b1a2bba7b70c2b52a7c5bea443929d787a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR