4-Butylaniline ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B0272-500ML B0272-25ML B0272-100ML
TCB0272-500MLEA 379.2 CAD
TCB0272-500ML TCB0272-25ML TCB0272-100ML
4-Butylaniline ≥97.0% (by GC, titration analysis)
4-Butylaniline

CAS Number: 104-13-2
MDL Number: MFCD00007925
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 250
Flash Point (°C): 101
Specific Gravity (20/20): 0.94

Formula: C₁₀H₁₅N
Boiling Pt: 250 °C
Melting Pt: –14 °C
Density: 0.94 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00007925
CAS Number: 104-13-2
EINECS: 203-177-4
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=093875d3ee1ae6a13c994693924097f217065904&vid=026aef81eb88ce8b2096e3433e388a81e7fa6f9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR