4-Phenylbenzaldehyde ≥95.0%

Supplier: TCI America
Warning

Synonyms: Biphenyl-4-carbaldehyde, [1,1'-Biphenyl]-4-carbaldehyde, 4-Biphenylcarboxaldehyde, Biphenyl-4-carboxaldehyde, p-Biphenylylaldehyde, 4-Biphenylylaldehyde, 4-Biphenylcarbaldehyde

B0242-250G B0242-25G B0242-5G
TCB0242-250GEA 641.48 CAD
TCB0242-250G TCB0242-025G TCB0242-005G
4-Phenylbenzaldehyde ≥95.0%
4-Phenylbenzaldehyde

CAS Number: 3218-36-8
MDL Number: MFCD00006947
Molecular Formula: C13H10O
Molecular Weight: 182.22
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 152
Melting point (°C): 58
Flash Point (°C): 93


Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction)
Other Product Names / Synonyms: 4-Phenylbenzaldehyde

Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 152 °C
Melting Pt: 58 °C
Flash Pt: 93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006947
CAS Number: 3218-36-8
EINECS: 221-742-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e7d459b8015024825feb966a21131c71e77608c&vid=b11ec2ab755f509ec022b684b409521c252ffec8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR