Benzyldimethylhexadecylammonium chloride ≥97.0% (by HPLC)

Supplier: TCI America

B0237-500G B0237-25G
TCB0237-500GEA 627.26 CAD
TCB0237-500G TCB0237-25G
Benzyldimethylhexadecylammonium chloride ≥97.0% (by HPLC)
Benzyldimethylhexadecylammonium chloride

CAS Number: 122-18-9
MDL Number: MFCD00011625
Molecular Formula: C25H46ClN
Molecular Weight: 396.10
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 65


Product Categories: Materials Chemistry, Cationic Surfactants
Other Product Names / Synonyms: Benzyldimethylhexadecylammonium Chloride, Benzylhexadecyldimethylammonium Chloride

Formula: C₂₅H₄₆ClN
Melting Pt: 65 °C
MDL Number: MFCD00011625
CAS Number: 122-18-9
EINECS: 204-526-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8348a129b257d87e0d6ba485db95e39de2e44b8e&vid=e185b133dbff435a1610fb4b384e02e2dfae8577&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR