Bromothymol blue 0.1% in ethanol 50%

Supplier: TCI America

Synonyms: 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide3′,3″-Dibromothymolsulfonphthalein3′,3″-Dibromothymolsulphonphthalein

B0113-500ML
TCB0113-500MLEA 150.51 CAD
TCB0113-500ML
Bromothymol blue 0.1% in ethanol 50%
Bromothymol blue

[for Titration]
CAS Number: 76-59-5
MDL Number: MFCD00005872
Molecular Weight: 624.38
Flash Point (°C): 24

Formula: C₂₇H₂₈Br₂O₅S
MW: 624.39 g/mol
Flash Pt: 24 °C
MDL Number: MFCD00005872
CAS Number: 76-59-5
ADR: 3,III
https://ca.vwr.com/stibo/web/std.lang.all/22/72/28932272.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR