2-Benzylphenol ≥97.0%
Supplier: TCI America
|
Warning
|
B0111-25G
B0111-500G
TCB0111-025GEA
67.94
CAD
TCB0111-025G
TCB0111-500G
2-Benzylphenol ≥97.0%
2-Benzylphenol
CAS Number: 28994-41-4
MDL Number: MFCD00002242
Molecular Formula: C13H12O
Molecular Weight: 184.24
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 312
Melting point (°C): 54
Flash Point (°C): 110
Other Product Names / Synonyms: 2-Hydroxydiphenylmethane, alpha-Phenyl-o-cresol
Formula:
C₁₃H₁₂O Boiling Pt: 312 °C Melting Pt: 54 °C Density: 1.1102 g/cm³ (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00002242 CAS Number: 28994-41-4 EINECS: 249-361-8 Merck Index: 13,01144 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...