Benzoylacetonitrile ≥98.0%
Supplier: TCI America
B0098-25G
B0098-5G
TCB0098-025GEA
143.78
CAD
TCB0098-025G
TCB0098-5G
Benzoylacetonitrile ≥98.0%
Benzoylacetonitrile
CAS Number: 614-16-4
MDL Number: MFCD00001942
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 81
Other Product Names / Synonyms: 2-Cyanoacetophenone
Formula:
(C₆H₅)C(O)CH₂CN Boiling Pt: 160 °C Melting Pt: 81 °C |
MDL Number:
MFCD00001942 CAS Number: 614-16-4 EINECS: 210-369-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...