Ethyl benzoylacetate ≥95.0%

Supplier: TCI America

Synonyms: Ethyl 3-oxo-3-phenylpropanoate

B0097-500G B0097-25G B0097-100G
TCB0097-500GEA 297.04 CAD
TCB0097-500G TCB0097-025G TCB0097-100G
Ethyl benzoylacetate ≥95.0%
Ethyl benzoylacetate

CAS Number: 94-02-0
MDL Number: MFCD00009196
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 270
Flash Point (°C): 141
Specific Gravity (20/20): 1.12


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Benzoylacetic Acid Ethyl Ester, Ethyl 3-Oxo-3-phenylpropionate, 3-Oxo-3-phenylpropionic Acid Ethyl Ester

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 265
Melting Pt: < 0 °C
Density: 1.12 (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00009196
CAS Number: 94-02-0
EINECS: 202-295-3
Merck Index: 13,03803
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=760630078853b4a2530c61216f72bfb3c8648d87&vid=760630078853b4a2530c61216f72bfb3c8648d87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR