2-Mercaptobenzoxazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Benzo[d]oxazole-2-thiol
B0096-25G
B0096-500G
TCB0096-025GEA
134.3
CAD
TCB0096-025G
TCB0096-500G
2-Mercaptobenzoxazole ≥98.0% (by GC, titration analysis)
2-Mercaptobenzoxazole
CAS Number: 2382-96-9
MDL Number: MFCD00005769
Molecular Formula: C7H5NOS
Molecular Weight: 151.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 196
Other Product Names / Synonyms: 2-Benzoxazolethiol
Formula:
C₇H₅NOS MW: 151.19 g/mol Melting Pt: 196 °C |
MDL Number:
MFCD00005769 CAS Number: 2382-96-9 EINECS: 219-191-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...