Benzotriazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Danger
|
B0094-500G
B0094-25G
B0094-100G
TCB0094-500GEA
150.1
CAD
TCB0094-500G
TCB0094-025G
TCB0094-100G
Benzotriazole ≥98.0% (by HPLC, titration analysis)
Benzotriazole
CAS Number: 95-14-7
MDL Number: MFCD00005699
Molecular Formula: C6H5N3
Molecular Weight: 119.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 204
Melting point (°C): 98
Flash Point (°C): 187
Other Product Names / Synonyms: Azimidobenzene
Formula:
C₆H₅N₃ Boiling Pt: 204 °C Melting Pt: 98 °C Density: 1.34 g/cm³ (20 °C) Flash Pt: 187 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005699 CAS Number: 95-14-7 EINECS: 202-394-1 UN: 2811 ADR: 6.1,III Merck Index: 13,01109 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...