Benzoin ≥98.0%

Supplier: TCI America

B0079-500G B0079-25G
TCB0079-500GEA 103.54 CAD
TCB0079-500G TCB0079-025G
Benzoin ≥98.0%
Benzoin

CAS Number: 119-53-9
MDL Number: MFCD00004496
Molecular Formula: C14H12O2
Molecular Weight: 212.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 344
Melting point (°C): 134


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: Benzoylphenylcarbinol, 2-Hydroxy-2-phenylacetophenone

Formula: C₁₄H₁₂O₂
Boiling Pt: 344 °C
Melting Pt: 134 °C
MDL Number: MFCD00004496
CAS Number: 119-53-9
EINECS: 204-331-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4630446783653e9e35a05fd1d6d43f79994bdda&vid=d4630446783653e9e35a05fd1d6d43f79994bdda&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR