Benzoic anhydride ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B0078-25G B0078-500G B0078-100G
TCB0078-025GEA 55.3 CAD
TCB0078-025G TCB0078-500G TCB0078-100G
Benzoic anhydride ≥97.0% (by GC, titration analysis)
Benzoic anhydride

CAS Number: 93-97-0
MDL Number: MFCD00003073
Molecular Formula: C14H10O3
Molecular Weight: 226.23
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 158
Melting point (°C): 42
Flash Point (°C): 150
Storage Temperature: 0-10°C

Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Boiling Pt: 158 °C
Melting Pt: 42 °C
Density: 1.231…1.247 g/cm³ (4 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00003073
CAS Number: 93-97-0
EINECS: 202-291-1
Merck Index: 13,01093
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b714d649aeb21aa728a7acf405e36be144c3a37&vid=8b714d649aeb21aa728a7acf405e36be144c3a37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR