Phenyl benzoate ≥99.0%

Supplier: TCI America

B0075-25G B0075-500G B0075-100G
TCB0075-025GEA 38.58 CAD
TCB0075-025G TCB0075-500G TCB0075-100G
Phenyl benzoate ≥99.0%
Phenyl benzoate

CAS Number: 93-99-2
MDL Number: MFCD00003072
Molecular Formula: C13H10O2
Molecular Weight: 198.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 314
Melting point (°C): 70


Other Product Names / Synonyms: Benzoic Acid Phenyl Ester

Formula: C₁₃H₁₀O₂
Boiling Pt: 314 °C
Melting Pt: 70 °C
Density: 1.041 g/cm³ (131 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003072
CAS Number: 93-99-2
EINECS: 202-293-2
Merck Index: 12,07429
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa563876f30e6b77f3400ae9db16d30ad295799d&vid=8d83020541f03364b4b563b86efa0e52f93bf862&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR