Benzoic acid methyl ester ≥99.0%

Supplier: TCI America
Warning

Synonyms: Methyl benzoate

B0074-25ML B0074-500ML
TCB0074-025MLEA 36.34 CAD
TCB0074-025ML TCB0074-500ML
Benzoic acid methyl ester ≥99.0%
Benzoic acid methyl ester

CAS Number: 93-58-3
MDL Number: MFCD00008421
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Melting point (°C): -12
Flash Point (°C): 91
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: Benzoic Acid Methyl Ester

Formula: C₆H₅COOCH₃
MW: 136.15 g/mol
Boiling Pt: 199.6
Melting Pt: -12
Density: 1.09 (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00008421
CAS Number: 93-58-3
EINECS: 202-259-7
Merck Index: 13,06052
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88112e38a7a258f851dd72a5e649852cbc3be3af&vid=aed1097251c094bc08b73b1e1100afee5e262304&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR