Isobutyl benzoate ≥99.0%

Supplier: TCI America

Synonyms: Benzoic acid isobutyl ester

B0072-500G B0072-25G
TCB0072-500GEA 284.4 CAD
TCB0072-500G TCB0072-025G
Isobutyl benzoate ≥99.0%
Isobutyl benzoate

CAS Number: 120-50-3
MDL Number: MFCD00048344
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 237
Flash Point (°C): 120
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Benzoic Acid Isobutyl Ester

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 237 °C
Density: 1.00 (20 °C)
Flash Pt: 120 °C
MDL Number: MFCD00048344
CAS Number: 120-50-3
EINECS: 204-401-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fb3e28f5e5b7d5fb03f4b6915e3f8dc5307bb84&vid=4d269d19b9b8ee183fc5bc512231bf901a70d93c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR