Ethyl benzoate ≥99.0%
Supplier: TCI America
|
Warning
|
B0069-25ML
B0069-500ML
TCB0069-025MLEA
30.02
CAD
TCB0069-025ML
TCB0069-500ML
Ethyl benzoate ≥99.0%
Ethyl benzoate
CAS Number: 93-89-0
MDL Number: MFCD00009109
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 212
Melting point (°C): -34
Flash Point (°C): 99
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: Benzoic Acid Ethyl Ester
Formula:
C₉H₁₀O₂ Boiling Pt: 212 °C Melting Pt: –34 °C Density: 1.05 (20 °C) Flash Pt: 99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009109 CAS Number: 93-89-0 EINECS: 202-284-3 Merck Index: 13,03802 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...