Butyl benzoate ≥99.0%

Supplier: TCI America
Warning

B0066-500ML B0066-25ML
TCB0066-500MLEA 69.09 CAD
TCB0066-500ML TCB0066-025ML
Butyl benzoate ≥99.0%
Butyl benzoate

CAS Number: 136-60-7
MDL Number: MFCD00009439
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 250
Melting point (°C): -22
Flash Point (°C): 122
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Benzoic Acid Butyl Ester

Formula: C₁₁H₁₄O₂
Boiling Pt: 250 °C
Melting Pt: –22 °C
Density: 1.01 (20 °C)
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00009439
CAS Number: 136-60-7
EINECS: 205-252-7
Merck Index: 13,01551
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a20f8114911663e5081ff5019d2b660699a04b7e&vid=da540721aa4cf4533f92aa52b258b06bee811390&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR