Benzyl benzoate ≥99.0%

Supplier: TCI America
Warning

Synonyms: Benzoic acid benzyl ester

B0064-25G B0064-500G
TCB0064-025GEA 33.76 CAD
TCB0064-025G TCB0064-500G
Benzyl benzoate ≥99.0%
Benzyl benzoate

CAS Number: 120-51-4
MDL Number: MFCD00003075
Molecular Formula: C14H12O2
Molecular Weight: 212.25
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 324
Flash Point (°C): 158
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: Benzoic Acid Benzyl Ester

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 324 °C
Melting Pt: 17…20 °C
Density: 1.12 (20 °C)
Flash Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD00003075
CAS Number: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9 III
Merck Index: 13,01129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a2d91d51e392603adc8dab1f1ad87c50804db18&vid=8f3ba19cbd3861bef124085b153272fa3d7a0e28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR