α-Naphthoflavone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-Phenyl-4H-benzo[h]chromen-4-one, 2-Phenylbenzo[h]chromen-4-one, 7,8-Benzoflavone

B0056-1G B0056-5G
TCB0056-001GEA 30.02 CAD
TCB0056-001G TCB0056-005G
α-Naphthoflavone ≥98.0% (by HPLC)
α-Naphthoflavone

CAS Number: 604-59-1
MDL Number: MFCD00004985
Molecular Formula: C19H12O2
Molecular Weight: 272.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 156


Other Product Names / Synonyms: 7,8-Benzoflavone

Formula: C₁₉H₁₂O₂
MW: 272.3 g/mol
Boiling Pt: 431 °C (1013 hPa)
Melting Pt: 156 °C
MDL Number: MFCD00004985
CAS Number: 604-59-1
EINECS: 210-071-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b0d8f57f49bcc9ecd98e4fc474595a45c4c2a1f&vid=8b0d8f57f49bcc9ecd98e4fc474595a45c4c2a1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR