Benzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1H-Benzo[d]imidazole, 1H-Benzimidazole

B0054-500G B0054-25G B0054-100G
TCB0054-500GEA 225.94 CAD
TCB0054-500G TCB0054-025G TCB0054-100G
Benzimidazole ≥98.0% (by GC, titration analysis)
Benzimidazole

CAS Number: 51-17-2
MDL Number: MFCD00005585
Molecular Formula: C7H6N2
Molecular Weight: 118.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 173
Lambda max.: 271 nm (MeOH)

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 173 °C
Density: 1.23 g/cm³ (20 °C)
MDL Number: MFCD00005585
CAS Number: 51-17-2
EINECS: 200-081-4
Merck Index: 13,01082
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70a396813b41826af2d705fbe09f8d7d09fc22d5&vid=70a396813b41826af2d705fbe09f8d7d09fc22d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR